missing translation for 'onlineSavingsMsg'
Learn More

[5-(Trifluoromethyl)-2-furyl]methylamine, 97%, Maybridge

Brand:  Maybridge MO00181DA

Code : 31

Additional Details : CAS Number : 868755-68-4 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 8 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10380414

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

[5-(Trifluoromethyl)-2-furyl]methylamine
97.0
97%
165.11
868755-68-4
97%
C6H6F3NO
MFCD03086222

Specifications

[5-(Trifluoromethyl)-2-furyl]methylamine
868755-68-4
Amber Glass Bottle
97%
165.11
1g
97.0
97%
C6H6F3NO
MFCD03086222
Videos
SDS