[5-(Trifluoromethyl)-2-furyl]methanol, ≥97%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: CC49809DA

1GR [5-(Trifluoromethyl)-2-furyl]methanol, 97%

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 65865-28-3 Weight: 0.00100kg Transport: UN number : 1986 Chem class : 1 Pack group : III


Packaging Amber Glass Bottle
Quantity 1g

Product Code. 10311153

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Chemical Identifiers

Chemical Name or Material [5-(Trifluoromethyl)-2-furyl]methanol
CAS 65865-28-3
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H5F3O2
Formula Weight 166.1
MDL Number MFCD08060542

Specification

Formula Weight 166.1
Quantity 1g
Assay 97%
CAS Min % 96.5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 65865-28-3
Assay Percent Range 97%
MDL Number MFCD08060542
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.
Packaging Amber Glass Bottle
Molecular Formula C6H5F3O2
Health Hazard 1 GHS Signal Word: Warning
CAS Max % 100.0
Chemical Name or Material [5-(Trifluoromethyl)-2-furyl]methanol
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Warning

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.