missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)-2-furaldehyde, ≥97%, Thermo Scientific™

Catalog Number p-3176240
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
17515-80-9
C6H3F3O2
164.083
MFCD00525585
KLVCIHFXPHKSPD-UHFFFAOYSA-N
5-trifluoromethyl furan-2-carbaldehyde, 5-trifluoromethyl-2-furaldehyde, 2-furancarboxaldehyde, 5-trifluoromethyl, 2-furancarboxaldehyde,5-trifluoromethyl, 5-trifluoromethyl furfural, 2-formyl-5-trifluoromethyl furan, 5-trifluoromethyl-furan-2-carbaldehyde, 5-trifluoromethyl-2-furancarboxaldehyde, 5-trifluoromethyl furan-2-carboxaldehyde
7537665
5-(trifluoromethyl)furan-2-carbaldehyde
C1=C(OC(=C1)C(F)(F)F)C=O
This item is not returnable. View return policy
17515-80-9
C6H3F3O2
164.083
MFCD00525585
KLVCIHFXPHKSPD-UHFFFAOYSA-N
5-trifluoromethyl furan-2-carbaldehyde, 5-trifluoromethyl-2-furaldehyde, 2-furancarboxaldehyde, 5-trifluoromethyl, 2-furancarboxaldehyde,5-trifluoromethyl, 5-trifluoromethyl furfural, 2-formyl-5-trifluoromethyl furan, 5-trifluoromethyl-furan-2-carbaldehyde, 5-trifluoromethyl-2-furancarboxaldehyde, 5-trifluoromethyl furan-2-carboxaldehyde
7537665
5-(trifluoromethyl)furan-2-carbaldehyde
C1=C(OC(=C1)C(F)(F)F)C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 164.08
Percent Purity 97%
Chemical Name or Material 5-(Trifluoromethyl)-2-furaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'