missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Trifluoromethoxy)-1H-indole-2-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 250 mg |
Formula Weight | 245.16 |
Percent Purity | 97% |
Chemical Name or Material | 5-(Trifluoromethoxy)-1H-indole-2-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'