Learn More
5-Thien-2-yl-1h-pyrazole-3-carboxylic acid, Maybridge™
Brand: Maybridge CC43501DE
Code : 31
Additional Details : CAS Number : 182415-24-3 Weight : 0.05500kg
Quantity | 5g |
---|
Chemical Identifiers
182415-24-3 | |
194.208 | |
DUDBNCNNJMOLCC-UHFFFAOYSA-N | |
818268 | |
C1=CSC(=C1)C2=CC(=NN2)C(=O)O |
C8H6N2O2S | |
MFCD05170066 | |
5-thiophen-2-yl-1h-pyrazole-3-carboxylic acid, 5-thiophen-2-yl-2h-pyrazole-3-carboxylic acid, 3-2-thienyl-1h-pyrazole-5-carboxylic acid, 5-thien-2-yl-1h-pyrazole-3-carboxylic acid, 5-2-thienyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-2-thienyl, 5-2-thienyl-3-pyrazolecarboxylic acid, 3-thiophen-2-yl-1h-pyrazole-5-carboxylic acid, 5-2-thienyl pyrazole-3-carboxylic acid | |
5-thiophen-2-yl-1H-pyrazole-3-carboxylic acid |
Specifications
5-Thien-2-yl-1H-pyrazole-3-carboxylic acid | |
182415-24-3 | |
100.0 | |
C8H6N2O2S | |
5-thiophen-2-yl-1h-pyrazole-3-carboxylic acid, 5-thiophen-2-yl-2h-pyrazole-3-carboxylic acid, 3-2-thienyl-1h-pyrazole-5-carboxylic acid, 5-thien-2-yl-1h-pyrazole-3-carboxylic acid, 5-2-thienyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-2-thienyl, 5-2-thienyl-3-pyrazolecarboxylic acid, 3-thiophen-2-yl-1h-pyrazole-5-carboxylic acid, 5-2-thienyl pyrazole-3-carboxylic acid | |
C1=CSC(=C1)C2=CC(=NN2)C(=O)O | |
194.208 | |
194.21 |
5g | |
96.5 | |
97% | |
MFCD05170066 | |
DUDBNCNNJMOLCC-UHFFFAOYSA-N | |
5-thiophen-2-yl-1H-pyrazole-3-carboxylic acid | |
818268 | |
97% |