missing translation for 'onlineSavingsMsg'
Learn More

(5-Pyrrolidin-1-ylpyrid-2-yl)methanol, 97%, Thermo Scientific™

Catalog Number p-3541165
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
930110-98-8
C10H14N2O
178.24
MFCD09702371
GLWULQJNDPJFDR-UHFFFAOYSA-N
5-pyrrolidin-1-ylpyrid-2-yl methanol, 5-pyrrolidin-1-ylpyridin-2-yl methanol, 5-pyrrolidin-1-yl pyridin-2-yl methanol, 5-pyrrolidinyl-2-pyridyl methan-1-ol, 5-pyrrolidine-1yl-pyrid-2-yl methanol, 2-pyridinemethanol,5-1-pyrrolidinyl, 1-6-hydroxymethyl pyridin-3-yl pyrrolidine, 2-hydroxymethyl-5-pyrrolidin-1-yl pyridine
24229532
OCC1=NC=C(C=C1)N1CCCC1
This item is not returnable. View return policy
930110-98-8
C10H14N2O
178.24
MFCD09702371
GLWULQJNDPJFDR-UHFFFAOYSA-N
5-pyrrolidin-1-ylpyrid-2-yl methanol, 5-pyrrolidin-1-ylpyridin-2-yl methanol, 5-pyrrolidin-1-yl pyridin-2-yl methanol, 5-pyrrolidinyl-2-pyridyl methan-1-ol, 5-pyrrolidine-1yl-pyrid-2-yl methanol, 2-pyridinemethanol,5-1-pyrrolidinyl, 1-6-hydroxymethyl pyridin-3-yl pyrrolidine, 2-hydroxymethyl-5-pyrrolidin-1-yl pyridine
24229532
OCC1=NC=C(C=C1)N1CCCC1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name [5-(pyrrolidin-1-yl)pyridin-2-yl]methanol
Formula Weight 178.23
Percent Purity 95%
Chemical Name or Material (5-Pyrrolidin-1-ylpyrid-2-yl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'