missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrimidinecarboxylic acid, 97%, Maybridge

Brand:  Maybridge CC07401DE

Code : 31

Additional Details : CAS Number : 4595-61-3 Weight : 0.05500kg

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10749195

  • £240.00 / 5g


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Chemical Identifiers

5-Pyrimidinecarboxylic acid
97%
124.1
97%
C5H4N2O2

Specifications

5-Pyrimidinecarboxylic acid
4595-61-3
97%
C5H4N2O2
5g
Amber Glass Bottle
97%
124.1
Videos
SDS