missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Pyridin-3-ylthiophene-2-carbaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS | 133531-43-8 |
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C10H7NOS |
MDL Number | MFCD00115001 |
Quantity | 1 g |
Synonym | 5-pyridin-3-yl thiophene-2-carbaldehyde, 5-3-pyridinyl-2-thiophenecarbaldehyde, 5-pyridin-3-yl-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-3-pyridinyl, acmc-20muz5, 5-3-pyridyl thiophene-2-carbaldehyde |
InChI Key | DIQWXENHCURRML-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'