missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Pyrid-4-ylthiophene-2-carbaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 5-pyridin-4-yl thiophene-2-carbaldehyde, 5-pyrid-4-ylthiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-4-pyridinyl, acmc-20mtde, 4-5-formylthien-2-yl pyridine, 2-formyl-5-pyridin-4-yl thiophene, 5-4-pyridyl thiophene-2-carbaldehyde, 5-4-pyridinyl-2-thiophenecarboxaldehyde, 2-thiophenecarboxaldehyde, 5-4-pyridinyl |
Formula Weight | 189.24 |
Percent Purity | 97% |
Chemical Name or Material | 5-Pyrid-4-ylthiophene-2-carbaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'