missing translation for 'onlineSavingsMsg'
Learn More

(5-Pyrid-3-ylthien-2-yl)methylamine, 97%, Thermo Scientific™

Catalog Number p-3586545
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
837376-47-3
C10H10N2S
190.264
MFCD09879956
JHLIGYPHPBLDDL-UHFFFAOYSA-N
5-pyrid-3-ylthien-2-yl methylamine, 5-pyridin-3-yl thiophen-2-yl methanamine, 1-5-pyridin-3-yl thiophen-2-yl methanamine, 5-pyridin-3-ylthiophen-2-yl methanamine, nicotine 3-heteroaromatic analogue 1a, 5-3-pyridyl-2-thienyl methylamine, 5-3-pyridyl-2-thienyl methanamine, compound 38a pmid: 15634016, 2-thiophenemethanamine,5-3-pyridinyl, 2-thiophenemethanamine, 5-3-pyridinyl
11206290
(5-pyridin-3-ylthiophen-2-yl)methanamine
C1=CC(=CN=C1)C2=CC=C(S2)CN
This item is not returnable. View return policy
837376-47-3
C10H10N2S
190.264
MFCD09879956
JHLIGYPHPBLDDL-UHFFFAOYSA-N
5-pyrid-3-ylthien-2-yl methylamine, 5-pyridin-3-yl thiophen-2-yl methanamine, 1-5-pyridin-3-yl thiophen-2-yl methanamine, 5-pyridin-3-ylthiophen-2-yl methanamine, nicotine 3-heteroaromatic analogue 1a, 5-3-pyridyl-2-thienyl methylamine, 5-3-pyridyl-2-thienyl methanamine, compound 38a pmid: 15634016, 2-thiophenemethanamine,5-3-pyridinyl, 2-thiophenemethanamine, 5-3-pyridinyl
11206290
(5-pyridin-3-ylthiophen-2-yl)methanamine
C1=CC(=CN=C1)C2=CC=C(S2)CN
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 190.27
Percent Purity 95%
Chemical Name or Material (5-Pyrid-3-ylthien-2-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'