missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Phenyltetrazole, 99%, Thermo Scientific Chemicals
14.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Quantity | 5 g |
Solubility Information | Solubility in water: .. Other solubilities: soluble in organic solvents |
Formula Weight | 146.15 |
Percent Purity | 99% |
Show More |
Safety and Handling
Product Identifier
- 5-Phenyltetrazole
- Warning
- Acute toxicity Category 4
- Flammable solid Category 2
- H302-Harmful if swallowed.
- H228-Flammable solid.
- EUH044-Risk of explosion if heated under confinement.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'