missing translation for 'onlineSavingsMsg'
Learn More

(5-Phenylisoxazol-3-yl)methanol, 97%, Thermo Scientific™

Catalog Number p-3484503
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
1619-37-0
C10H9NO2
175.187
MFCD01444104
BPAFLGGUEBMWRN-UHFFFAOYSA-N
1481082
(5-phenyl-1,2-oxazol-3-yl)methanol
C1=CC=C(C=C1)C2=CC(=NO2)CO
This item is not returnable. View return policy
1619-37-0
C10H9NO2
175.187
MFCD01444104
BPAFLGGUEBMWRN-UHFFFAOYSA-N
1481082
(5-phenyl-1,2-oxazol-3-yl)methanol
C1=CC=C(C=C1)C2=CC(=NO2)CO
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 5-phenylisoxazol-3-yl methanol, 5-phenyl-3-isoxazolyl methanol, 5-phenyl-1,2-oxazol-3-yl methanol, 3-isoxazolemethanol, 5-phenyl, 3-hydroxymethyl-5-phenylisoxazole, 5-phenylisoxazol-3-yl methan-1-ol, 5-phenylisoxazole-3-methanol, 3-isoxazolemethanol,5-phenyl, 5-phenyl-3-hydroxymethylisoxazole, 5-phenyl-isoxazol-3-yl methanol
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material (5-Phenylisoxazol-3-yl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'