missing translation for 'onlineSavingsMsg'
Learn More

(5-Phenyl-1,3-oxazol-4-yl)methanol, ≥95%, Thermo Scientific™

Catalog Number p-2693784
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
352018-88-3
C10H9NO2
175.187
MFCD03086105
LVHWTAMRDRDXJP-UHFFFAOYSA-N
5-phenyloxazol-4-yl methanol, 5-phenyl-1,3-oxazol-4-yl methanol, 5-phenyl-1,3-oxazole-4-methanol, 4-oxazolemethanol,5-phenyl, 5-phenyl-4-oxazolyl methanol, 5-phenyl-oxazol-4-yl-methanol, 5-phenyl-1,3-oxazol-4-yl methan-1-ol
2776302
(5-phenyl-1,3-oxazol-4-yl)methanol
C1=CC=C(C=C1)C2=C(N=CO2)CO
This item is not returnable. View return policy
352018-88-3
C10H9NO2
175.187
MFCD03086105
LVHWTAMRDRDXJP-UHFFFAOYSA-N
5-phenyloxazol-4-yl methanol, 5-phenyl-1,3-oxazol-4-yl methanol, 5-phenyl-1,3-oxazole-4-methanol, 4-oxazolemethanol,5-phenyl, 5-phenyl-4-oxazolyl methanol, 5-phenyl-oxazol-4-yl-methanol, 5-phenyl-1,3-oxazol-4-yl methan-1-ol
2776302
(5-phenyl-1,3-oxazol-4-yl)methanol
C1=CC=C(C=C1)C2=C(N=CO2)CO
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 175.19
Percent Purity 95%
Chemical Name or Material (5-Phenyl-1, 3-oxazol-4-yl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'