missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Oxo-1-(2-thienylmethyl)pyrrolidine-3-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Synonym | 5-oxo-1-2-thienylmethyl pyrrolidine-3-carboxylic acid, 5-oxo-1-thiophen-2-ylmethyl pyrrolidine-3-carboxylic acid, 4-carboxy-1-then-2-yl pyrrolidin-2-one, 3-pyrrolidinecarboxylicacid, 5-oxo-1-2-thienylmethyl, 5-oxo-1-thien-2-ylmethyl pyrrolidine-3-carboxylic acid, maybridge1_003341, 3-pyrrolidinecarboxylicacid,5-oxo-1-2-thienylmethyl, 5-oxo-1-2-thienylmethyl-3-pyrrolidinecarboxylic acid, 5-oxo-1-2-thienylmethyl-3-pyrrolidinecarboxylic acid #, 5-oxo-1-thiophen-2-ylmethyl-pyrrolidine-3-ca rboxylic acid |
Formula Weight | 225.27 |
Percent Purity | 97% |
Chemical Name or Material | 5-Oxo-1-(2-thienylmethyl)pyrrolidine-3-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'