missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-carboxaldehyde, 95%, mixture of endo and exo, Thermo Scientific Chemicals

Catalog Number p-175613
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5453-80-5
C8H10O
122.17
MFCD00167568
AJIBZRIAUXVGQJ-UHFFFAOYSA-N
5-norbornene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carbaldehyde, 2-formyl-5-norbornene, 5-formylbicyclohept-2-ene, 5-norbornene-2-carbaldehyde, 5-norbornane-2-carboxaldehyde, bicyclo 2.2.1 hept-2-ene-5-carbaldehyde
95117
bicyclo[2.2.1]hept-2-ene-5-carbaldehyde
C1C2CC(C1C=C2)C=O
This item is not returnable. View return policy
5453-80-5
C8H10O
122.17
MFCD00167568
AJIBZRIAUXVGQJ-UHFFFAOYSA-N
5-norbornene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carbaldehyde, 2-formyl-5-norbornene, 5-formylbicyclohept-2-ene, 5-norbornene-2-carbaldehyde, 5-norbornane-2-carboxaldehyde, bicyclo 2.2.1 hept-2-ene-5-carbaldehyde
95117
bicyclo[2.2.1]hept-2-ene-5-carbaldehyde
C1C2CC(C1C=C2)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 90.0
CAS Max % 100.0
Density 1.0180g/mL
Boiling Point 67°C to 70°C (12.0 mmHg)
Flash Point 50.6°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4870 to 1.492
Quantity 5 g
Specific Gravity 1.018
Solubility Information Solubility in water: insoluble
Formula Weight 122.17
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5-Norbornene-2-carboxaldehyde, 90-95%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Norbornene-2-carboxaldehyde, mixture of endo and exo
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'