Learn More
5-Nonyloxytryptamine oxalate, Tocris Bioscience™
Selective 5-HT1B agonist
Brand:  Tocris (Bio-Techne) 090150
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13428133
  • £610.05 / 50mg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
5-HT1B selective agonist, several times more potent than sumatriptan and inactive as a 5-HT1A agonist (Ki at 5-HT1B = 1 nM, selectivity over 5-HT1A > 300-fold). Mimics polysialic acid activity, stimulates neuritogenesis, myelination and Schwann cell migration in vitro.
Specifications
Chemical Identifiers
157798-13-5
392.496
5-nonyloxytryptamine oxalate, nonyloxytryptamine oxalate, 2-5-nonyloxy-1h-indol-3-yl ethanamine ethanedioate, 2-5-nonyloxy-1h-indol-3-yl ethanamine oxalate, o-nonylserotonin oxalate, 3-2-aminoethyl-5-nonyloxyindole oxalate, 5-nonyloxy-1h-indole-3-ethanamine hydrogen maleate, 2-5-nonyloxy-1h-indol-3-yl ethanaminium hydrogen oxalate
CHEBI:64147
CCCCCCCCCOC1=CC2=C(C=C1)NC=C2CCN.C(=O)(C(=O)O)O
C21H32N2O5
JORSCLBFSAAOFR-UHFFFAOYSA-N
10126593
2-(5-nonoxy-1H-indol-3-yl)ethanamine;oxalic acid
Specifications
50mg
157798-13-5
5-nonyloxytryptamine oxalate, nonyloxytryptamine oxalate, 2-5-nonyloxy-1h-indol-3-yl ethanamine ethanedioate, 2-5-nonyloxy-1h-indol-3-yl ethanamine oxalate, o-nonylserotonin oxalate, 3-2-aminoethyl-5-nonyloxyindole oxalate, 5-nonyloxy-1h-indole-3-ethanamine hydrogen maleate, 2-5-nonyloxy-1h-indol-3-yl ethanaminium hydrogen oxalate
CCCCCCCCCOC1=CC2=C(C=C1)NC=C2CCN.C(=O)(C(=O)O)O
392.496
CHEBI:64147
>99%
5-Nonyloxytryptamine oxalate
C21H32N2O5
JORSCLBFSAAOFR-UHFFFAOYSA-N
2-(5-nonoxy-1H-indol-3-yl)ethanamine;oxalic acid
10126593
392.49