missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrouracil, 99+%, ACROS Organics™

Brand:  Acros Organics 226580050

Code : 29

Additional Details : CAS Number : 611-08-5 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10519433

  • £9.40 / 5g


Estimated Shipment
15-06-2018
Add to basket


Specifications

Chemical Identifiers

5-Nitrouracil
611-08-5
100.0
99+%
157.08
p.a.
99.0
≥99%
C4H3N3O4
MFCD00006021

Specifications

5-Nitrouracil
611-08-5
100.0
Glass bottle
Off-white to pale yellow
≥99%
5g
157.08
24, 320
p.a.
99.0
Authentic
> 300.0°C
Crystalline Powder or Needles
99+%
C4H3N3O4
MFCD00006021
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Nitrouracil

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS