missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrothiophene-3-carboxylic acid, 97%, Maybridge

£26.00 - £194.00

Chemical Identifiers

CAS 40357-96-8
Molecular Formula C5H3NO4S
Molecular Weight (g/mol) 173.142
MDL Number MFCD00052582
InChI Key VNJUNWUOAKEIKG-UHFFFAOYSA-N
Synonym 5-nitrothiophene-3-carboxylic acid, 2-nitrothiophene-4-carboxylic acid, 5-nitro-thiophene-3-carboxylic acid, 5-nitro-3-thiophenecarboxylic acid, 3-thiophenecarboxylic acid, 5-nitro, maybridge1_003249, acmc-1ascr, 2-nitro-4-carboxythiophene, ksc238c5f, 2-nitro-4-thiophenecarboxylic acid
PubChem CID 2777267
SMILES C1=C(SC=C1C(=O)O)[N+](=O)[O-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10009441 SDS View Documents Maybridge
DFP00046CB
250mg £26.00
250mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 14-05-2020
Please to purchase this item. Need a web account? Register with us today!
10653213 SDS View Documents Maybridge
DFP00046DA
1g £70.00
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 14-05-2020
Please to purchase this item. Need a web account? Register with us today!
10182744 SDS View Documents Maybridge
DFP00046DE
5g £194.00
5g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 14-05-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

40357-96-8
173.142
VNJUNWUOAKEIKG-UHFFFAOYSA-N
2777267
C5H3NO4S
MFCD00052582
5-nitrothiophene-3-carboxylic acid, 2-nitrothiophene-4-carboxylic acid, 5-nitro-thiophene-3-carboxylic acid, 5-nitro-3-thiophenecarboxylic acid, 3-thiophenecarboxylic acid, 5-nitro, maybridge1_003249, acmc-1ascr, 2-nitro-4-carboxythiophene, ksc238c5f, 2-nitro-4-thiophenecarboxylic acid
C1=C(SC=C1C(=O)O)[N+](=O)[O-]

Specifications

5-Nitrothiophene-3-carboxylic acid
250mg
96.5
97%
MFCD00052582
VNJUNWUOAKEIKG-UHFFFAOYSA-N
173.142
173.15
Amber Glass Bottle
40357-96-8
100.0
C5H3NO4S
5-nitrothiophene-3-carboxylic acid, 2-nitrothiophene-4-carboxylic acid, 5-nitro-thiophene-3-carboxylic acid, 5-nitro-3-thiophenecarboxylic acid, 3-thiophenecarboxylic acid, 5-nitro, maybridge1_003249, acmc-1ascr, 2-nitro-4-carboxythiophene, ksc238c5f, 2-nitro-4-thiophenecarboxylic acid
C1=C(SC=C1C(=O)O)[N+](=O)[O-]
2777267
97%