missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitrothiophene-3-carbonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS | 42137-23-5 |
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C5H2N2O2S |
MDL Number | MFCD00052580 |
Quantity | 1 g |
Synonym | 2-nitrothiophene-4-carbonitrile, acmc-1aogv, maybridge1_003117, 2-nitro-4-cyanothiophene, 4-cyano-2-nitrothiophene, 5-nitro-3-thiophenecarbonitrile, nacmzlgghmpmkq-uhfffaoysa, 5-nitro-3-thiophenecarbonitrile #, 5-nitro-thiophene-3-carbonitrile, 5-nitro-3-thiophenecarbonitrile; 2-nitro-4-cyanothiophene; 2-cyano-4-nitrothiophene |
InChI Key | NACMZLGGHMPMKQ-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'