missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrothiophene-2-carbonitrile, 98%, Thermo Scientific™

Catalog Number p-4813234
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
16689-02-4
C5H2N2O2S
154.15
FLYONFCGDKAMIH-UHFFFAOYSA-N
5-nitro-2-thiophenecarbonitrile, 5-cyano-2-nitrothiophene, 2-cyano-5-nitrothiophene, 5-nitrothiophene-2-carbonitirile, 5-nitro-thiophene-2-carbonitrile, 2-thiophenecarbonitrile, 5-nitro, zlchem 478, maybridge1_005980, 5-nitro-2-cyanothiophene, acmc-209dv8
519294
5-nitrothiophene-2-carbonitrile
C1=C(SC(=C1)[N+](=O)[O-])C#N
This item is not returnable. View return policy
16689-02-4
C5H2N2O2S
154.15
FLYONFCGDKAMIH-UHFFFAOYSA-N
5-nitro-2-thiophenecarbonitrile, 5-cyano-2-nitrothiophene, 2-cyano-5-nitrothiophene, 5-nitrothiophene-2-carbonitirile, 5-nitro-thiophene-2-carbonitrile, 2-thiophenecarbonitrile, 5-nitro, zlchem 478, maybridge1_005980, 5-nitro-2-cyanothiophene, acmc-209dv8
519294
5-nitrothiophene-2-carbonitrile
C1=C(SC(=C1)[N+](=O)[O-])C#N
Melting Point 42°C to 46°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 154.15
Percent Purity 98%
Chemical Name or Material 5-Nitrothiophene-2-carbonitrile
compliance-icons
Product Identifier
  • 5-Nitro-2-thiophenecarbonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'