missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrosalicylic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-175092
Click to view available options
Quantity:
5 g
25 g
250 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
96-97-9
C7H5NO5
183.12
MFCD00007338
PPDRLQLKHRZIJC-UHFFFAOYSA-N
5-nitrosalicylic acid, 2-hydroxy-5-nitrobenzoic acid, anilotic acid, benzoic acid, 2-hydroxy-5-nitro, 5-nitro-2-hydroxybenzoic acid, unii-82l9g7fyz3, 2-hydroxy-5-nitro-benzoic acid, 5-nitro-salicylic acid, salicylic acid, 5-nitro, 5-nitro salicylic acid
7318
CHEBI:61281
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)O
This item is not returnable. View return policy
96-97-9
C7H5NO5
183.12
MFCD00007338
PPDRLQLKHRZIJC-UHFFFAOYSA-N
5-nitrosalicylic acid, 2-hydroxy-5-nitrobenzoic acid, anilotic acid, benzoic acid, 2-hydroxy-5-nitro, 5-nitro-2-hydroxybenzoic acid, unii-82l9g7fyz3, 2-hydroxy-5-nitro-benzoic acid, 5-nitro-salicylic acid, salicylic acid, 5-nitro, 5-nitro salicylic acid
7318
CHEBI:61281
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 228°C to 231°C
Color Beige-Yellow to Brown-Orange
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula O2NC6H3(OH)COOH
Quantity 5 g
Beilstein 10, 116
Merck Index 15, 6718
Solubility Information Solubility in water: 1g/1475mL in water. Other solubilities: more soluble in hot water,very soluble in ethanol,ether,acetone
Formula Weight 183.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 5-Nitrosalicylic acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'