missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitroisophthalic acid, 97+%, Thermo Scientific Chemicals
12.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Description
5-Nitroisophthalic acid is used as an intermediate of medicine compound and as a disperser in dyes. It is used as an intermediate in organic synthesis,agrochemical,pharmaceutical and dyestuffs etc,.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Nitroisophthalic acid is used as an intermediate of medicine compound and as a disperser in dyes. It is used as an intermediate in organic synthesis,agrochemical,pharmaceutical and dyestuffs etc,.
Solubility
Soluble in water.1.5 g/L at 20°C
Notes
Incompatible with strong oxidizing agents, strong bases.
5-Nitroisophthalic acid is used as an intermediate of medicine compound and as a disperser in dyes. It is used as an intermediate in organic synthesis,agrochemical,pharmaceutical and dyestuffs etc,.
Solubility
Soluble in water.1.5 g/L at 20°C
Notes
Incompatible with strong oxidizing agents, strong bases.
Specifications
Melting Point | 260°C to 264°C |
Odor | Odorless |
Quantity | 100 g |
Beilstein | 1976587 |
Solubility Information | Soluble in water.1.5g/L at 20°C |
Formula Weight | 211.13 |
Percent Purity | 98% |
Chemical Name or Material | 5-Nitroisophthalic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'