missing translation for 'onlineSavingsMsg'
Learn More

5-nitrohexahydropyrimidine-2,4,6-trione hydrate, 97%, Thermo Scientific™

Catalog Number p-3485928
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
175278-58-7
C4H5N3O6
191.10
MFCD00160499
YKGZAIJTCJOZNK-UHFFFAOYSA-N
5-nitropyrimidine-2,4,6 1h,3h,5h-trione xhydrate, 5-nitrohexahydropyrimidine-2,4,6-trione hydrate, 5-nitro-1,3-diazinane-2,4,6-trione hydrate, 5-nitropyrimidine-2,4,6 1h,3h,5h-trione hydrate, 5-nitrobarbituric acid hydrate, c4nh33o5.h2o, 5-nitro-hexahydro-pyrimidine-2,4,6-trione, 2,4,6 1h,3h,5h-pyrimidinetrione,5-nitro-, hydrate 1:1
2781274
O.[O-][N+](=O)C1C(=O)NC(=O)NC1=O
This item is not returnable. View return policy
175278-58-7
C4H5N3O6
191.10
MFCD00160499
YKGZAIJTCJOZNK-UHFFFAOYSA-N
5-nitropyrimidine-2,4,6 1h,3h,5h-trione xhydrate, 5-nitrohexahydropyrimidine-2,4,6-trione hydrate, 5-nitro-1,3-diazinane-2,4,6-trione hydrate, 5-nitropyrimidine-2,4,6 1h,3h,5h-trione hydrate, 5-nitrobarbituric acid hydrate, c4nh33o5.h2o, 5-nitro-hexahydro-pyrimidine-2,4,6-trione, 2,4,6 1h,3h,5h-pyrimidinetrione,5-nitro-, hydrate 1:1
2781274
O.[O-][N+](=O)C1C(=O)NC(=O)NC1=O
CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
IUPAC Name 5-nitro-1,3-diazinane-2,4,6-trione hydrate
Formula Weight 191.1
Percent Purity 97%
Chemical Name or Material 5-Nitrohexahydropyrimidine-2, 4, 6-trione hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'