missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Nitro-3-(1-pyrrolidinylmethyl)indole , 95%

Brand:  Alfa Aesar™ H63372.03

Code : NEW

Additional Details : CAS Number : 3414-68-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15410039

  • £95.61 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

5-Nitro-3-(1-pyrrolidinylmethyl)indole
245.28
421075
3414-68-4
MFCD01718616

Specifications

5-Nitro-3-(1-pyrrolidinylmethyl)indole
Solid
1g
MFCD01718616
3414-68-4
95%
245.28
421075
Videos
SDS