missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Nitro-3-(1-piperidinylmethyl)indole, 95%

Brand:  Alfa Aesar™ H63195.03

Code : NEW

Additional Details : CAS Number : 3414-69-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449758

  • £95.61 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

5-Nitro-3-(1-piperidinylmethyl)indole
259.3
425671
3414-69-5
MFCD01718615

Specifications

5-Nitro-3-(1-piperidinylmethyl)indole
Solid
259.3
1g
3414-69-5
95%
MFCD01718615
425671
Videos
SDS