missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Nitro-3-(1,2,3,6-tetrahydro-4-pyridyl)indole, 97%

Brand:  Alfa Aesar™ H63134.06

Code : NEW

Additional Details : CAS Number : 109793-85-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15478136

  • £478.06 / 5g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

5-Nitro-3-(1,2,3,6-tetrahydro-4-pyridyl)indole
243.26
4193644
109793-85-3
MFCD26741987

Specifications

5-Nitro-3-(1,2,3,6-tetrahydro-4-pyridyl)indole
109793-85-3
97%
MFCD26741987
5g
Solid
243.26
4193644
Videos
SDS