missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Nitro-2-phenylbenzimidazole, 95%

Brand:  Alfa Aesar™ H64772.03

Code : NEW

Additional Details : CAS Number : 1571-85-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15412119

  • £119.78 / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

5-Nitro-2-phenylbenzimidazole
1571-85-3
239.23
MFCD00225448

Specifications

5-Nitro-2-phenylbenzimidazole
Solid
239.23
1g
1571-85-3
95%
MFCD00225448
Videos
SDS