missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Nitro-2-phenylbenzimidazole, 95%

Brand:  Alfa Aesar™ H64772.06

Code : NEW

Additional Details : CAS Number : 1571-85-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15422119

  • £597.84 / 5g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

5-Nitro-2-phenylbenzimidazole
1571-85-3
239.23
MFCD00225448

Specifications

5-Nitro-2-phenylbenzimidazole
Solid
239.23
5g
1571-85-3
95%
MFCD00225448
Videos
SDS