missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitro-2-furaldehyde, 97%, Thermo Scientific Chemicals
92.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber glass bottle
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 34.0°C to 35.0°C |
Color | Yellow to Brown |
Density | 1.3400g/mL |
Flash Point | 33°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C5H3NO4 |
Show More |
Safety and Handling
Product Identifier
- 5-Nitro-2-furaldehyde
- Warning
- Flammable solid Category 2
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'