missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furaldehyde, 97%, ACROS Organics™

Brand:  Maybridge AC17386DA

Code : 31

Additional Details : CAS Number : 698-63-5 Weight : 0.05100kg Transport : UN number : 1325 Chem class : 1 Pack group : III

Quantity 1g
Packaging Amber glass bottle

 View more versions of this product

Product Code. 10518663

  • £7.00 / 1g


Estimated Shipment date
from Supplier 25-01-2018
Add to basket

Specifications

Chemical Identifiers

5-Nitro-2-furaldehyde
96.0
≥96.0%
C5H3NO4
MFCD00003230
698-63-5
100.0
97%
141.08
5-Nitrofurfural

Specifications

5-Nitro-2-furaldehyde
Yellow to brown
1g
96.0
100.0
Crystalline Low Melting Solid
97%
141.08
5-Nitrofurfural
1.3400g/mL
33° to 37°C
698-63-5
Authentic
Amber glass bottle
≥96.0%
C5H3NO4
MFCD00003230
Videos
SDS