missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Nitro-2-(2-thienyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64885.06

Code : NEW

Additional Details : CAS Number : 37128-73-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15412289

  • £398.56 / 5g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

5-Nitro-2-(2-thienyl)benzimidazole
37128-73-7
245.26
MFCD00441576

Specifications

5-Nitro-2-(2-thienyl)benzimidazole
37128-73-7
95%
MFCD00441576
5g
Solid
245.26
Videos
SDS