missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-1H-pyrazole, 90%, Thermo Scientific™

Catalog Number p-3485926
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
26621-44-3
C3H3N3O2
113.08
MFCD00159621,MFCD00238787
MZRUFMBFIKGOAL-UHFFFAOYSA-N
3-nitro-1h-pyrazole, 3-nitropyrazole, 1h-pyrazole, 3-nitro, 3-nitro-2h-pyrazole, nitropyrazole, pyrazole, 3-nitro, 5-nitro-1h-pyrazol, djw 94, djw-94, 5-nitropyrazole
123419
5-nitro-1H-pyrazole
[O-][N+](=O)C1=CC=NN1
This item is not returnable. View return policy
26621-44-3
C3H3N3O2
113.08
MFCD00159621,MFCD00238787
MZRUFMBFIKGOAL-UHFFFAOYSA-N
3-nitro-1h-pyrazole, 3-nitropyrazole, 1h-pyrazole, 3-nitro, 3-nitro-2h-pyrazole, nitropyrazole, pyrazole, 3-nitro, 5-nitro-1h-pyrazol, djw 94, djw-94, 5-nitropyrazole
123419
5-nitro-1H-pyrazole
[O-][N+](=O)C1=CC=NN1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 113.08
Percent Purity 97%
Chemical Name or Material 5-Nitro-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'