Learn More
Thermo Scientific Maybridge 5-Nitro-1H-indole, 25g, 97%
Brand: Thermo Scientific Maybridge SB01605EB
5-Nitro-1H-indole, 25g, 97%
Code : 31
Additional Details : CAS Number : 6146-52-7 Weight : 0.07500kg
Brand: Thermo Scientific Maybridge SB01605EB
5-Nitro-1H-indole, 25g, 97%
Code : 31
Additional Details : CAS Number : 6146-52-7 Weight : 0.07500kg