missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Nitro-1-indoleacetic acid, 95%

Brand:  Alfa Aesar™ H63759.03

Code : NEW

Additional Details : CAS Number : 226901-50-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15490609

  • £95.61 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

5-Nitro-1-indoleacetic acid
220.18
11128841
226901-50-4
MFCD11205787

Specifications

5-Nitro-1-indoleacetic acid
226901-50-4
95%
MFCD11205787
1g
Solid
220.18
11128841
Videos
SDS