missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-2-carboxaldehyde, 98+%, Thermo Scientific™

Catalog Number p-7081199
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
13679-70-4
C6H6OS
126.173
MFCD00005434
VAUMDUIUEPIGHM-UHFFFAOYSA-N
5-methyl-2-thiophenecarboxaldehyde, 5-methylthiophene-2-carboxaldehyde, 5-methyl-2-thiophenecarbaldehyde, 2-formyl-5-methylthiophene, 5-methyl-2-formylthiophene, 2-thiophenecarboxaldehyde, 5-methyl, 5-methylthiophene-2-aldehyde, 5-methyl-2-thenaldehyde, 5-methyl-2-thiophencarboxaldehyde, 5-methyl-2-thiophene carboxaldehyde
61663
5-methylthiophene-2-carbaldehyde
CC1=CC=C(S1)C=O
This item is not returnable. View return policy
13679-70-4
C6H6OS
126.173
MFCD00005434
VAUMDUIUEPIGHM-UHFFFAOYSA-N
5-methyl-2-thiophenecarboxaldehyde, 5-methylthiophene-2-carboxaldehyde, 5-methyl-2-thiophenecarbaldehyde, 2-formyl-5-methylthiophene, 5-methyl-2-formylthiophene, 2-thiophenecarboxaldehyde, 5-methyl, 5-methylthiophene-2-aldehyde, 5-methyl-2-thenaldehyde, 5-methyl-2-thiophencarboxaldehyde, 5-methyl-2-thiophene carboxaldehyde
61663
5-methylthiophene-2-carbaldehyde
CC1=CC=C(S1)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Methylthiophene-2-carboxaldehyde is used as a food additive. It is an analytical reagent. The reagent has been synthesized and characterized using IR, 1H NMR and Mass spectral data.

Solubility
Soluble in ether and ethanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Density 1.18
Boiling Point 92°C to 93°C (12 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.583
Quantity 10 g
Beilstein 106896
Sensitivity Air Sensitive
Solubility Information Soluble in ether and ethanol.
Formula Weight 126.18
Percent Purity ≥98%
Chemical Name or Material 5-Methylthiophene-2-carboxaldehyde
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'