missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Methylthiophene-2-carboxaldehyde, 98+%

Brand:  Alfa Aesar™ A11119.18

Code : NEW

Additional Details : CAS Number : 13679-70-4 Weight : 0.05000kg

Quantity 50g

 View more versions of this product

Product Code. 11419013

  • £107.00 / 50g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Description

Description

Applications
5-Methylthiophene-2-carboxaldehyde is used as a food additive. It is an analytical reagent. The reagent has been synthesized and characterized using IR, 1H NMR and Mass spectral data.

Solubility
Soluble in ether and ethanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

5-Methylthiophene-2-carboxaldehyde
C6H6OS
MFCD00005434
13679-70-4
126.18
106896

Specifications

5-Methylthiophene-2-carboxaldehyde
≥98%
126.18
106896
50g
13679-70-4
C6H6OS
MFCD00005434
92° to 93°C (12mmHg)
87°C (188°F)
Videos
SDS