missing translation for 'onlineSavingsMsg'
Learn More

5-(Methylthio)thiophene-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number 15436276
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
20873-58-9
C6H6O2S2
174.232
MFCD00173742
FWFWUSLBIIIIEN-UHFFFAOYSA-N
5-methylthio thiophene-2-carboxylic acid, 5-methylsulfanyl thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 5-methylthio, 2-thiophenecarboxylicacid, 5-methylthio, 5-methylthiothiophene-2-carboxylic acid, maybridge1_007717, d04ngc, cambridge id 5103856, 5-methylmercaptothiophene-2-carboxylic, 5-methylthio-2-thiophenecarboxylic acid
714372
5-methylsulfanylthiophene-2-carboxylic acid
CSC1=CC=C(S1)C(=O)O
This item is not returnable. View return policy
20873-58-9
C6H6O2S2
174.232
MFCD00173742
FWFWUSLBIIIIEN-UHFFFAOYSA-N
5-methylthio thiophene-2-carboxylic acid, 5-methylsulfanyl thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 5-methylthio, 2-thiophenecarboxylicacid, 5-methylthio, 5-methylthiothiophene-2-carboxylic acid, maybridge1_007717, d04ngc, cambridge id 5103856, 5-methylmercaptothiophene-2-carboxylic, 5-methylthio-2-thiophenecarboxylic acid
714372
5-methylsulfanylthiophene-2-carboxylic acid
CSC1=CC=C(S1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 103°C to 105°C
Color White
Quantity 250 mg
Formula Weight 174.24
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-(Methylthio)thiophene-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'