missing translation for 'onlineSavingsMsg'
Learn More

5-(Methylsulfonyl)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog Number 11946471
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1206641-26-0
C12H18BNO4S
283.15
MFCD12198145
BFWBUXKEBXAKDE-UHFFFAOYSA-N
3-methylsulfonyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 5-methylsulfonyl pyridine-3-boronic acid pinacol ester, 3-methanesulfonyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-methanesulfonyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-methylsulfonyl pyridine-5-boronic acid pinacol ester, 5-methylsulfonyl pyridine-3-boronic acid, pinacol ester, 5-methylsulfonyl pyridin-3-ylboronic acid pinacol ester, 3-methylsulfonyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 3-methylsulfonyl-5-4,4,5,5-tetramethyl-1,3,2-dioxab oro lan-2-yl pyridine
53398530
3-methylsulfonyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1(C)OB(OC1(C)C)C1=CC(=CN=C1)S(C)(=O)=O
This item is not returnable. View return policy
1206641-26-0
C12H18BNO4S
283.15
MFCD12198145
BFWBUXKEBXAKDE-UHFFFAOYSA-N
3-methylsulfonyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 5-methylsulfonyl pyridine-3-boronic acid pinacol ester, 3-methanesulfonyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-methanesulfonyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-methylsulfonyl pyridine-5-boronic acid pinacol ester, 5-methylsulfonyl pyridine-3-boronic acid, pinacol ester, 5-methylsulfonyl pyridin-3-ylboronic acid pinacol ester, 3-methylsulfonyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 3-methylsulfonyl-5-4,4,5,5-tetramethyl-1,3,2-dioxab oro lan-2-yl pyridine
53398530
3-methylsulfonyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1(C)OB(OC1(C)C)C1=CC(=CN=C1)S(C)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 164°C to 167°C
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 283.15
Percent Purity 95%
Chemical Name or Material 5-(Methylsulfonyl)pyridine-3-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 5-(Methylsulfonyl)pyridine-3-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'