missing translation for 'onlineSavingsMsg'
Learn More

5-Methylpyrimidine, 98%, Thermo Scientific™

Catalog Number 15264844
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2036-41-1
C5H6N2
94.12
MFCD00047386
TWGNOYAGHYUFFR-UHFFFAOYSA-N
pyrimidine, 5-methyl, 5-methyl-pyrimidine, pyrimidine, 5-methyl-6ci,7ci,8ci,9ci, 5-methyl-pyrimidin, pubchem7207, rarechem am la 0031, 5-methyl-1,3-diazine, acmc-1cnd2, 5-methylpyrimidine, 5-methylpyrimidine, 97+%
74859
5-methylpyrimidine
CC1=CN=CN=C1
This item is not returnable. View return policy
2036-41-1
C5H6N2
94.12
MFCD00047386
TWGNOYAGHYUFFR-UHFFFAOYSA-N
pyrimidine, 5-methyl, 5-methyl-pyrimidine, pyrimidine, 5-methyl-6ci,7ci,8ci,9ci, 5-methyl-pyrimidin, pubchem7207, rarechem am la 0031, 5-methyl-1,3-diazine, acmc-1cnd2, 5-methylpyrimidine, 5-methylpyrimidine, 97+%
74859
5-methylpyrimidine
CC1=CN=CN=C1
Melting Point 33°C to 37°C
Boiling Point 152°C to 154°C
Flash Point 52°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble
Formula Weight 94.11
Percent Purity 98%
Chemical Name or Material 5-Methylpyrimidine
compliance-icons compliance-icons
Product Identifier
  • 5-Methylpyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'