missing translation for 'onlineSavingsMsg'
Learn More

5-Methylpyridine-2-carbonitrile, ≥97%, Thermo Scientific Chemicals

Catalog Number 15421809
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1620-77-5
C7H6N2
118.14
MFCD06200830
LIEQVZZZYLHNRH-UHFFFAOYSA-N
2-cyano-5-methylpyridine, 5-methylpicolinonitrile, 5-methyl-pyridine-2-carbonitrile, 5-methyl-2-pyridinecarbonitrile, 6-cyano-3-picoline, 2-pyridinecarbonitrile, 5-methyl, 2-cyano-5-methylpyridine 2-cyano-5-picoline, 5-methypicolinonitrile, pubchem12978, 5-methyl picolinonitrile
74183
5-methylpyridine-2-carbonitrile
CC1=CC=C(N=C1)C#N
This item is not returnable. View return policy
1620-77-5
C7H6N2
118.14
MFCD06200830
LIEQVZZZYLHNRH-UHFFFAOYSA-N
2-cyano-5-methylpyridine, 5-methylpicolinonitrile, 5-methyl-pyridine-2-carbonitrile, 5-methyl-2-pyridinecarbonitrile, 6-cyano-3-picoline, 2-pyridinecarbonitrile, 5-methyl, 2-cyano-5-methylpyridine 2-cyano-5-picoline, 5-methypicolinonitrile, pubchem12978, 5-methyl picolinonitrile
74183
5-methylpyridine-2-carbonitrile
CC1=CC=C(N=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 72°C to 75°C
Quantity 1 g
UN Number UN3439
Beilstein 111125
Formula Weight 118.14
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 5-Methylpyridine-2-carbonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'