missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Methylpyridine-2-carbonitrile, ≥97%

Brand:  Alfa Aesar™ H64539.14

Code : NEW

Additional Details : CAS Number : 1620-77-5 Weight : 0.02500kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15441809

  • £207.70 / 25g


Estimated Shipment date
from Supplier 24-04-2018
Add to basket

Specifications

Chemical Identifiers

5-Methylpyridine-2-carbonitrile
118.14
UN3439
5-Methylpicolinonitrile; 2-Cyano-5-methylpyridine
1620-77-5
MFCD06200830
111125

Specifications

5-Methylpyridine-2-carbonitrile
1620-77-5
≥97%
MFCD06200830
111125
5-Methylpicolinonitrile; 2-Cyano-5-methylpyridine
25g
Solid
118.14
UN3439
72° to 75°C
Videos
SDS