missing translation for 'onlineSavingsMsg'
Learn More

5-Methylpyridine-2-boronic acid N-phenyldiethanolamine ester, 50-70%, ACROS Organics™

Brand:  Acros Organics 439950010

Code : 29

Additional Details : CAS Number : 872054-54-1 Weight : 0.05100kg Transport : UN number : 3175 Chem class : 1 Pack group : II


Quantity 1g

 View more versions of this product

Product Code. 11906011

  • £43.00 / 1g


Estimated Shipment
23-05-2018
Add to basket


Specifications

Chemical Identifiers

5-Methylpyridine-2-boronic acid N-phenyldiethanolamine ester
C16H19BN2O2
2-(5-Methyl-pyridin-2-yl)-6-phenyl-(1,3,6,2)dioxazaborolane
872054-54-1
282.15

Specifications

5-Methylpyridine-2-boronic acid N-phenyldiethanolamine ester
872054-54-1
282.15
2-(5-Methyl-pyridin-2-yl)-6-phenyl-(1,3,6,2)dioxazaborolane
1g
C16H19BN2O2
> 300.0°C
Videos
SDS