missing translation for 'onlineSavingsMsg'
Learn More

5-Methylpyrazin-2-ylamine, 97%, Thermo Scientific™

Catalog Number 10547284
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
5521-58-4
C5H7N3
109.132
MFCD08437664
ZNQOALAKPLGUPH-UHFFFAOYSA-N
2-amino-5-methylpyrazine, 5-methyl-pyrazin-2-ylamine, 5-methyl-2-pyrazinamine, pyrazinamine, 5-methyl, 2-amino-5 methylpyrazine, 5-methyl-2-aminopyrazine, 2-amino-5-methyl pyrazine, 5-methylpyrazine-2-ylamine, 2-pyrazinamine, 5-methyl, pubchem15710
313215
5-methylpyrazin-2-amine
CC1=CN=C(C=N1)N
This item is not returnable. View return policy
5521-58-4
C5H7N3
109.132
MFCD08437664
ZNQOALAKPLGUPH-UHFFFAOYSA-N
2-amino-5-methylpyrazine, 5-methyl-pyrazin-2-ylamine, 5-methyl-2-pyrazinamine, pyrazinamine, 5-methyl, 2-amino-5 methylpyrazine, 5-methyl-2-aminopyrazine, 2-amino-5-methyl pyrazine, 5-methylpyrazine-2-ylamine, 2-pyrazinamine, 5-methyl, pubchem15710
313215
5-methylpyrazin-2-amine
CC1=CN=C(C=N1)N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 109.13
Percent Purity 97%
Chemical Name or Material 5-Methylpyrazin-2-ylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'