missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Methylisoxazole-3-carboxamide, 95%

Brand:  Alfa Aesar™ H63986.03

Code : NEW

Additional Details : CAS Number : 3445-52-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15410989

  • £149.46 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

5-Methylisoxazole-3-carboxamide
3445-52-1
126.11
MFCD00085128

Specifications

5-Methylisoxazole-3-carboxamide
Solid
126.11
1g
3445-52-1
95%
MFCD00085128
Videos
SDS