missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisatin, 95%, Thermo Scientific Chemicals

Catalog Number p-7077196
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
608-05-9
C9H7NO2
161.16
MFCD00005721
VAJCSPZKMVQIAP-UHFFFAOYSA-N
5-methylisatin, 5-methylindoline-2,3-dione, 5-methylindole-2,3-dione, 1h-indole-2,3-dione, 5-methyl, indole-2,3-dione, 5-methyl, 5-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-methyl-2,3-indolinedione, 5-methyl-indole-2,3-dione, 5-methylindole-2,3 1h-dione, 5-methyl-1h-benzo d azolidine-2,3-dione
11840
5-methyl-1H-indole-2,3-dione
CC1=CC=C2NC(=O)C(=O)C2=C1
This item is not returnable. View return policy
608-05-9
C9H7NO2
161.16
MFCD00005721
VAJCSPZKMVQIAP-UHFFFAOYSA-N
5-methylisatin, 5-methylindoline-2,3-dione, 5-methylindole-2,3-dione, 1h-indole-2,3-dione, 5-methyl, indole-2,3-dione, 5-methyl, 5-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-methyl-2,3-indolinedione, 5-methyl-indole-2,3-dione, 5-methylindole-2,3 1h-dione, 5-methyl-1h-benzo d azolidine-2,3-dione
11840
5-methyl-1H-indole-2,3-dione
CC1=CC=C2NC(=O)C(=O)C2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 180.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Beilstein 21,509
Solubility Information Solubility in water: insoluble.
Formula Weight 161.16
Percent Purity 95%
Chemical Name or Material 5-Methylisatin
compliance-icons
Product Identifier
  • 5-Methylisatin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'