missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylindole-2-carboxylic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
5-Methylindole-2-carboxylic acid is a compound used in the synthesis of Pin1 inhibitors as potential antitumor agents. reactant in preparation of Pin1 inhibitors as potential antitumor agents reactant in preparation of non-imidazole human histamine H4 receptor antagonists
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Methylindole-2-carboxylic acid is a compound used in the synthesis of Pin1 inhibitors as potential antitumor agents. reactant in preparation of Pin1 inhibitors as potential antitumor agents reactant in preparation of non-imidazole human histamine H4 receptor antagonists
Solubility
Soluble in ethanol (50 mg/ml). Insoluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
5-Methylindole-2-carboxylic acid is a compound used in the synthesis of Pin1 inhibitors as potential antitumor agents. reactant in preparation of Pin1 inhibitors as potential antitumor agents reactant in preparation of non-imidazole human histamine H4 receptor antagonists
Solubility
Soluble in ethanol (50 mg/ml). Insoluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Melting Point | ∽235°C (decomposition) |
Quantity | 1 g |
Beilstein | 144055 |
Solubility Information | Soluble in ethanol (50mg/ml). Insoluble in water. |
Formula Weight | 175.19 |
Percent Purity | 99% |
Chemical Name or Material | 5-Methylindole-2-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'