missing translation for 'onlineSavingsMsg'
Learn More

5-Methylfurfural, 98+%, Thermo Scientific Chemicals

Catalog Number p-163204
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
620-02-0
C6H6O2
110.11
MFCD00003232
OUDFNZMQXZILJD-UHFFFAOYSA-N
5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran
12097
CHEBI:2091
5-methylfuran-2-carbaldehyde
CC1=CC=C(O1)C=O
This item is not returnable. View return policy
620-02-0
C6H6O2
110.11
MFCD00003232
OUDFNZMQXZILJD-UHFFFAOYSA-N
5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran
12097
CHEBI:2091
5-methylfuran-2-carbaldehyde
CC1=CC=C(O1)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 10mg KOH/g max.
CAS Min % 98.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.1000g/mL
Boiling Point 187°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5290 to 1.532
Quantity 25 g
Beilstein 17, 289
Specific Gravity 1.1
Solubility Information Solubility in water: 33.3g/L (20 c)
Formula Weight 110.11
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 5-Methylfurfural, 98+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'