missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-phenyl-4-isoxazolesulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-3175633
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
857283-56-8
C10H8ClNO3S
257.688
MFCD06658966
GXQBTNFENYVPFM-UHFFFAOYSA-N
5-methyl-3-phenyl-4-isoxazolesulfonyl chloride, 5-methyl-3-phenylisoxazole-4-sulphonyl chloride, 5-methyl-3-phenylisoxazole-4-sulfonyl chloride, chloro 5-methyl-3-phenylisoxazol-4-yl sulfone, 5-methyl-3-phenyl-isoxazole-4-sulfonyl chloride
18525696
5-methyl-3-phenyl-1,2-oxazole-4-sulfonyl chloride
CC1=C(C(=NO1)C2=CC=CC=C2)S(=O)(=O)Cl
This item is not returnable. View return policy
857283-56-8
C10H8ClNO3S
257.688
MFCD06658966
GXQBTNFENYVPFM-UHFFFAOYSA-N
5-methyl-3-phenyl-4-isoxazolesulfonyl chloride, 5-methyl-3-phenylisoxazole-4-sulphonyl chloride, 5-methyl-3-phenylisoxazole-4-sulfonyl chloride, chloro 5-methyl-3-phenylisoxazol-4-yl sulfone, 5-methyl-3-phenyl-isoxazole-4-sulfonyl chloride
18525696
5-methyl-3-phenyl-1,2-oxazole-4-sulfonyl chloride
CC1=C(C(=NO1)C2=CC=CC=C2)S(=O)(=O)Cl
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 257.7
Percent Purity 97%
Chemical Name or Material 5-Methyl-3-phenyl-4-isoxazolesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'