missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5-Methyl-3-isoxazolyl)methylamine, 97+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 5-methyl-3-isoxazolyl methylamine, 5-methylisoxazol-3-yl methanamine, 5-methyl-1,2-oxazol-3-yl methanamine, 5-methylisoxazol-3-yl methylamine, 1-5-methyl-1,2-oxazol-3-yl methanamine, 1-5-methylisoxazol-3-yl methanamine, 3-isoxazolemethanamine,5-methyl, 3-isoxazolemethanamine, 5-methyl, c-5-methyl-isoxazol-3-yl-methylamine, acmc-20a4az |
Formula Weight | 112.13 |
Percent Purity | 97% |
Chemical Name or Material | (5-Methyl-3-isoxazolyl)methylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'