missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-(4-methylphenyl)-1,2,4-oxadiazole, 97%, Thermo Scientific™

Catalog Number p-7036150
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
81386-30-3
C10H10N2O
174.203
MFCD00085801
OYGSEQYNSZOSCE-UHFFFAOYSA-N
5-methyl-3-p-tolyl-1,2,4-oxadiazole, 5-methyl-3-4-methylphenyl-1,2,4-oxadiazole, 3-4-methylphenyl-5-methyl-1,2,4-oxadiazole, acmc-209pld, 5-methyl-3-4-methylphenyl-1,2,4-oxadiazole #
270750
5-methyl-3-(4-methylphenyl)-1,2,4-oxadiazole
CC1=CC=C(C=C1)C2=NOC(=N2)C
This item is not returnable. View return policy
81386-30-3
C10H10N2O
174.203
MFCD00085801
OYGSEQYNSZOSCE-UHFFFAOYSA-N
5-methyl-3-p-tolyl-1,2,4-oxadiazole, 5-methyl-3-4-methylphenyl-1,2,4-oxadiazole, 3-4-methylphenyl-5-methyl-1,2,4-oxadiazole, acmc-209pld, 5-methyl-3-4-methylphenyl-1,2,4-oxadiazole #
270750
5-methyl-3-(4-methylphenyl)-1,2,4-oxadiazole
CC1=CC=C(C=C1)C2=NOC(=N2)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 7438
Formula Weight 174.2
Percent Purity 97%
Chemical Name or Material 5-Methyl-3-(4-methylphenyl)-1,2,4-oxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'